missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Human UCH-L1 ELISA Kit

Product Code. 17224876 Shop All Proteintech Products
Click to view available options
Gene ID (Entrez):
7345
Unit Size:
96 reactions
This item is not returnable. View return policy

Product Code. 17224876

Brand: Proteintech KE0021196T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ubiquitin C-terminal hydrolase 1 (UCHL1) , previously known as PGP9.5, is found primarily in neurons and neuroendocrine cells with a very high concentration in dendrites and perikarya.

UCHL1 is a more recently recognized biomarker of neuronal injury. UCHL1 could be detected in the CSF and serum with a half-life of 7-9 hours after severe traumatic brain injury. Increased plasma concentrations of UCHL1 therefore reflect increased blood brain barrier permeability and destruction of neurons.

Specifications

Gene ID (Entrez) 7345
compliance-icons compliance-icons
Product Identifier
  • 96RXN Human UCH-L1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.