missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human UBE2A (aa 27-54) Control Fragment Recombinant Protein

Product Code. 30212678
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212678

Brand: Invitrogen™ RP105648

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Multiple alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49459
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7319
Name Human UBE2A (aa 27-54) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias E2 ubiquitin-conjugating enzyme A; hhr6a; HR6A; Mhr6a; mp:zf637-2-000771; MRXS30; MRXSN; RAD6 homolog A; Rad6a; si:bz46j2.4; UBC2; ube2a; ube2a.L; Ubiquitin carrier protein A; ubiquitin conjugating enzyme E2 A; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2A L homeolog; ubiquitin conjugating enzyme E2A; ubiquitin-conjugating enzyme E2 A; ubiquitin-conjugating enzyme E2A; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme HR6A; ubiquitin-protein ligase A; wu:fa01h11; XELAEV_18038882mg
Common Name UBE2A
Gene Symbol UBE2A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence APSENNIMVWNAVIFGPEGTPFEDGTFK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.