missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human u-Plasminogen Activator (uPA)/Urokinase Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1310
This item is not returnable.
View return policy
Description
u-Plasminogen Activator/Urokinase Polyclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| u-Plasminogen Activator/Urokinase | |
| Polyclonal | |
| Unconjugated | |
| P00749 | |
| PLAU | |
| Mouse myeloma cell line NS0-derived recombinant human uPA Ser21-Leu432 Accession # P00749 | |
| 100 μg | |
| Primary | |
| Detects human u-Plasminogen Activator in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5328 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction