missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TWEAK R/TNFRSF12 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1199
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blot.
Specifications
| TWEAK R/TNFRSF12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NP84.1 | |
| TNFRSF12A | |
| E. coli-derived recombinant human TWEAK R/TNFRSF12 Glu28-Trp79 Accession # Q9NP84.1 | |
| 100 μg | |
| Primary | |
| Detects human TWEAK R/TNFRSF12 in direct ELISAs and Western blots. Shows approximately 40% cross-reactivity with recombinant mouse TWEAK R and no cross-reactivity with recombinant human (rh) OPG or rhTNF RI. | |
| Human | |
| Purified |
| Western Blot | |
| 314502 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, Fn14, FN14tumor necrosis factor receptor superfamily member 12A, TNFRSF12, TNFRSF12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51330 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction