missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TUT1 (aa 802-871) Control Fragment Recombinant Protein

Product Code. 30212442
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212442

Brand: Invitrogen™ RP107672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TUT1 is a nucleotidyl transferase that functions as both a terminal uridylyltransferase and a nuclear poly(A) polymerase. TUT1 specifically adds and removes nucleotides from the 3' end of small nuclear RNAs and select mRNAs and may function in controlling gene expression and cell proliferation. TUT1 specifically catalyzes uridylylation of U6 snRNA (RNU6; MIM 180692) and is essential for cell proliferation (Trippe et al., 2006 [PubMed 16790842]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H6E5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64852
Name Human TUT1 (aa 802-871) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700038E08Rik; FLJ21850; FLJ22267; FLJ22347; MGC131987; MGC149809; nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpha regulated-poly(A) polymerase; nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase; PAP-associated domain-containing 2; PAPD2; poly(A) polymerase associated domain containing 2; RBM21; RNA binding motif protein 21; RNA uridylyltransferase; RNA-binding motif protein 21; RNA-binding protein 21; speckle targeted PIP5K1A-regulated poly(A) polymerase; STARPAP; Star-PAP; terminal uridylyl transferase 1 U6 snRNA-specific; terminal uridylyl transferase 1, U6 snRNA-specific; Tut1; TUTase; TUTase 6; TUTase6; U6 snRNA-specific terminal uridylyltransferase 1; U6 snRNA-specific terminal uridylyltransferase 1; speckle targeted PIP5K1A-regulated poly(A) polymerase; U6 TUTase; U6-TUTase; up-regulated in lung cancer 6; URLC6
Common Name TUT1
Gene Symbol TUT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GWLATEAQVTQELKGLSGGEERPETEPLLSFVASVSPADRMLTVTPLQDPQGLFPDLHHFLQVFLPQAIR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.