missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TUSC3 (aa 41-77) Control Fragment Recombinant Protein

Product Code. 30204281
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204281

Brand: Invitrogen™ RP103752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62109 (PA5-62109. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13454
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7991
Name Human TUSC3 (aa 41-77) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU022242; BC003311; D8S1992; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TUSC3; M33; Magnesium uptake/transporter TUSC3; MGC13453; MRT22; MRT7; N33; Oligosaccharyl transferase subunit TUSC3; oligosaccharyltransferase 3 homolog A; OST3A; Protein N33; Putative prostate cancer tumor suppressor; tumor suppressor candidate 3; Tusc3
Common Name TUSC3
Gene Symbol TUSC3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GQKKKENLLAEKVEQLMEWSSRRSIFRMNGDKFRKFI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.