missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tumstatin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7546-SP
This item is not returnable.
View return policy
Description
Tumstatin/COL4A3 Monoclonal specifically detects Tumstatin/COL4A3 in Human samples. It is validated for Western Blot.
Specifications
| Tumstatin/COL4A3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| COL4A3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), Goodpasture antigen, tumstatin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1285 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 762102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COL4A3 | |
| E. coli-derived recombinant human Tumstatin | |
| 25 μg | |
| Primary | |
| Detects human Tumstatin in direct ELISAs. In direct ELISAs, less than 25% cross-reactivity with recombinant human Collagen IV alpha 1 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction