missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Tubby (NP_003311) VersaClone cDNA
Description
- The Human Tubby (NP_003311) VersaClone cDNA clone contains the complete ORF for Tubby, along with a Kozak consensus sequence for optimal translation initiation
- The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
- The Tubby insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
Specifications
Specifications
| Gene | hTubby |
| Accession Number | NP_003311 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Gene ID (Entrez) | 7275 |
| Content And Storage | Store the unopened product at -20 to -80°C |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?