missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TTLL6 (aa 714-827) Control Fragment Recombinant Protein

Product Code. 30211419
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30211419 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30211419 Supplier Invitrogen™ Supplier No. RP104130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (41%), Rat (41%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62599 (PA5-62599. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N841
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 284076
Name Human TTLL6 (aa 714-827) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4932418K24Rik; D11Moh43e; D11Moh44e; polyglutamylase; RGD1311922; t8130b59; TTL0.6; TTLL6; Ttll6-ps1; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase like 6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; tubulin--tyrosine ligase-like protein 6
Common Name TTLL6
Gene Symbol TTLL6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLKSDMNKPHLISELLTKLQLSGKLSFFPAHYNPKLGMNNLSQNPSLPGECHSRSDSSGEKRQLDVSSLLLQSPQSYNVTLRDLLVIATPAQLDPRPCRSHASAMRDPCMQDQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.