missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TTC21B (aa 336-427) Control Fragment Recombinant Protein

Product Code. 30205762
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205762

Brand: Invitrogen™ RP109336

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140036 (PA5-140036. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. TTC21B (tetratricopeptide repeat domain 21B), also known as THM1, is a 1,316 amino acid protein that contains 19 TPR repeats and belongs to the TTC21 family. Localizing to cytoplasm and cytoskeleton, TTC21B exists as two alternatively spliced isoforms and is thought to negatively regulate Shh signal transduction. TTC21B may also be involved in retrograde intraflagellar transport in cilia, and is encoded by a gene that maps to human chromosome 2q24.3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z4L5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79809
Name Human TTC21B (aa 336-427) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410066K11Rik; aln; ATD4; IFT139; IFT139B; Intraflagellar transport 139 homolog; JBTS11; Kiaa1992; line 158; mKIAA1992; Nbla10696; NPHP12; putative protein product of Nbla10696; RGD1565122; SRTD4; tetratricopeptide repeat domain 21 B; tetratricopeptide repeat protein 21 B; Tetratricopeptide repeat-containing hedgehog modulator 1; Thm1; TPR repeat protein 21 B; Ttc21b
Common Name TTC21B
Gene Symbol TTC21B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LQGRVKEALKWYKTAMTLDETSVSALVGFIQCQLIEGQLQDADQQLEFLNEIQQSIGKSAELIYLHAVLAMKKNKRQEEVINLLNDVLDTHF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.