missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSPYL2 (aa 390-527) Control Fragment Recombinant Protein

Product Code. 30200805
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200805

Brand: Invitrogen™ RP92259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (55%), Rat (55%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111195 (PA5-111195. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H2G4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64061
Name Human TSPYL2 (aa 390-527) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CINAP; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; DENTT; differentially expressed nucleolar TGF-beta1 target; differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; testis-specific Y-encoded-like protein 2; TSPX; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2
Common Name TSPYL2
Gene Symbol TSPYL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NPLRYYLRERGSRIKRKKQEMKKRKTRGRCEVVIMEDAPDYYAVEDIFSEISDIDETIHDIKISDFMETTDYFETTDNEITDINENICDSENPDHNEVPNNETTDNNESADDHETTDNNESADDNNENPEDNNKNTDD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.