missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSPAN33 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8405-SP
This item is not returnable.
View return policy
Description
TSPAN33 Monoclonal specifically detects TSPAN33 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| TSPAN33 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| NP_848657 | |
| TSPAN33 | |
| NS0 mouse myeloma cell line transfected with human TSPAN33 Met1-Tyr283 Accession # NP_848657 | |
| 25 μg | |
| Primary | |
| Detects HEK293 human embryonic kidney cell line transfected with human TSPAN33 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 545422 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hPen, PEN, Penumbra, proerythroblast new membrane, tetraspanin 33, tetraspanin-33, tspan-33 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 340348 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction