missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human TSP-1 Platinum ELISA Kit
For the Detection of human Thrombospondin-1 (TSP-1)
£451.00 - £3070.00
Specifications
Assay Sensitivity | 0.044ng/mL |
---|---|
Sample Type | Plasma (EDTA, Citrate, Heparin), Serum |
Target Species | Human |
For Use With (Application) | ELISA |
Synonym | Thrombospondin 1, THBS1, THBS, THBS-1, TSP, TSP-1, TSP1 |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human TSP-1
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15561657
|
Invitrogen™
BMS2100 |
96 Tests |
£451.00
96 tests |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
15571657
|
Invitrogen™
BMS2100TEN |
10 x 96 Tests |
£3070.00
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
0.044ng/mL | |
Human | |
Thrombospondin 1, THBS1, THBS, THBS-1, TSP, TSP-1, TSP1 |
Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
TSP-1 |