missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TSG Recombinant Protein, PeproTech®

Product Code. 17811173
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17811173

Brand: Gibco™ 120091MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TSG is a 198 amino acid, 22.2 kDa protein containing the BMP/TGF-beta binding portion of the full length TSG protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZX9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57045
Molecular Weight (g/mol) 22.2 kDa
Name Human TSG
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1810013J15Rik; 9030422N06Rik; AW552143; D17Ertd403e; PSEC0250; Tsg; Twg; twisted gastrulation BMP signaling modulator 1; twisted gastrulation homolog 1; twisted gastrulation homolog 1 (Drosophila); twisted gastrulation protein homolog 1; Twsg1
Common Name TSG
Gene Symbol Twsg1
Biological Activity Determined by its ability to neutralize BMP-6-induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED50 for this effect is ≤ 2.0 ug/ml of TSG.
Conjugate Unconjugated
Recombinant Recombinant
Sequence CNKALCASDV SKCLIQELCQ CRPGEGNCSC CKECMLCLGA LWDECCDCVG MCNPRNYSDT PPTSKSTVEE LHEPIPSLFR ALTEGDTQLN WNIVSFPVAE ELSHHENLVS FLETVNQPHH QNVSVPSNNV HAPYSSDKEH MCTVVYFDDC MSIHQCKISC ESMGASKYRW FHNACCECIG PECIDYGSKT VKCMNCMF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.