missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TSEN2 (aa 93-189) Control Fragment Recombinant Protein

Product Code. 30205268
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205268

Brand: Invitrogen™ RP94795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (45%), Rat (45%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55513 (PA5-55513. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Constitutes one of the two catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5'- and 3'-splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2',3'-cyclic phosphate and 5'-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. Isoform 1 probably carries the active site for 5'-splice site cleavage. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre- mRNA 3'-end processing factors, establishing a link between pre- tRNA splicing and pre-mRNA 3'-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events. Isoform 2 is responsible for processing a yet unknown RNA substrate. The complex containing isoform 2 is not able to cleave pre-tRNAs properly, although it retains endonucleolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NCE0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80746
Name Human TSEN2 (aa 93-189) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU067695; HsSen2; PCH2B; RGD1309946; SEN2; SEN2L; tRNA splicing endonuclease 2 homolog (S. cerevisiae); tRNA splicing endonuclease subunit 2; tRNA-intron endonuclease Sen2; tRNA-intron nuclease 2; tRNA-splicing endonuclease subunit Sen2; Tsen2; TSEN2 tRNA splicing endonuclease subunit
Common Name TSEN2
Gene Symbol Tsen2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.