missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tryptophan Hydroxylase 1/TPH-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5276-SP
This item is not returnable.
View return policy
Description
Tryptophan Hydroxylase 1/TPH-1 Polyclonal specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPH-1, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7166 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P17752 | |
| TPH1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tryptophan Hydroxylase 1/TPH-1 Met102-Pro403 Accession # P17752 | |
| 25 μg | |
| Primary | |
| Detects human Tryptophan Hydroxylase 1/TPH-1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu