missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Tryptase alpha/TPS1 ELISA Kit (Chemiluminescence)

Product Code. 30279369 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30279369

Brand: Novus Biologicals™ NBP343199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemiluminescence ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.37 to 1000 pg/mL
Assay Sensitivity 0.61 pg/mL
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Sample Type Serum, Plasma, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7177
Gene Symbol TPSAB1
Kit Contents Pre-coated 96T Strip Plate, Plate sealer for 96 wells, Standard, Standard Diluent, Detection Reagent A, Assay Diluent A, Detection Reagent B, Assay Diluent B, Substrate A, Substrate B, Wash Buffer (30X concentrate), Instruction manual
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.