missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trypsin 3/PRSS3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3710
This item is not returnable.
View return policy
Description
Trypsin 3/PRSS3 Monoclonal specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for Neutralization.
Specifications
| Trypsin 3/PRSS3 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.15 ug/mL | |
| NP_002762 | |
| PRSS3 | |
| Mouse myeloma cell line NS0-derived recombinant human Trypsin 3/PRSS3 Val16-Ser247 Accession # NP_002762 | |
| 100 μg | |
| Primary | |
| Detects human Trypsin 3/PRSS3 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Trypsin-1 or rhTrypsin-2 is observed. | |
| Human | |
| Purified |
| Neutralization | |
| 420025 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsin, Mesotrypsinogen, MTG, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS3, PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin 4, Trypsin III, Trypsin IV, trypsin-3, Trypsinogen 15, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5646 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction