missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trypsin 3/PRSS3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB37101-SP
This item is not returnable.
View return policy
Description
Trypsin 3/PRSS3 Monoclonal specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for Western Blot.
Specifications
| Trypsin 3/PRSS3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_002762 | |
| PRSS3 | |
| Mouse myeloma cell line NS0-derived recombinant human Trypsin 3/PRSS3 Val16-Ser247 Accession # NP_002762 | |
| 25 μg | |
| Primary | |
| Detects human Trypsin 3/PRSS3 in direct ELISAs and Western blots. Detects both pro and mature forms of Trypsin 3 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Trypsin 1, 2, recombinant mouse Trypsin 3, or Chymostrypsin is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 420012 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsin, Mesotrypsinogen, MTG, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS3, PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin 4, Trypsin III, Trypsin IV, trypsin-3, Trypsinogen 15, trypsinogen 4, trypsinogen 5, trypsinogen IV | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5646 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction