missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Trypsin 2/PRSS2 ELISA Kit (Colorimetric)

Product Code. 18632061 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18632061

Brand: Novus Biologicals™ NBP2770541Kit

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.78 ng/mL to 50 ng/mL
Assay Sensitivity 0.19 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates
Specificity This assay has high sensitivity and excellent specificity for detection of Human Trypsin-2. No significant cross-reactivity or interference between Human Trypsin-2 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 5645
Gene Symbol PRSS2
Quantity 1 Kit
Research Discipline Cell Biology
Storage Requirements Store at 4°C.
Synonym Anionic trypsinogen, EC 3.4.21, EC 3.4.21.4, MGC111183, MGC120174, MGC120175, protease serine 2 preproprotein, protease, serine, 2 (trypsin 2), protease, serine, 2, preproprotein, Serine protease 2, TRY2TRY8, TRYP2, trypsin 2, Trypsin II, trypsin-2, trypsinogen 2
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.