missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Trypsin 1/PRSS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3848
This item is not returnable.
View return policy
Description
Trypsin 1/PRSS1 Monoclonal specifically detects Trypsin 1/PRSS1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Trypsin 1/PRSS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P07477 | |
| PRSS1 | |
| Mouse myeloma cell line NS0-derived recombinant human Trypsin 1/PRSS1 Ala16-Ser247 Accession # P07477 | |
| 100 μg | |
| Primary | |
| Detects human Trpysin 1/PRSS1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) Trypsin 3, recombinant human (rh) Trypsin 2, or rhTrypsin 3. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 446628 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), PRSS1, PTRYI, Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, Trygn16, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5644 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction