missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRPV2 (aa 27-108) Control Fragment Recombinant Protein

Product Code. 30181431
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181431

Brand: Invitrogen™ RP99981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

VLR-1 is a calcium-permeable, non-selective cation channel with an outward rectification, that seems to be regulated, at least in part, by growth factors, like IGF1, PDGF and morphogenetic neuropeptide/head activator. VRL-1 does not respond to capsaicin, acid, or moderate heat, but instead is activated by high temperatures, with a threshold of approximately 52 degrees C. Within sensory ganglia, VRL-1 is most prominently expressed by a subset of medium- to large-diameter neurons, making it a candidate receptor for transducing high-threshold heat responses in this class of cells. VRL-1 transcripts are not restricted to the sensory nervous system, indicating that this channel may be activated by stimuli other than heat. VRL-1 is one of the six members of the vanilloid-like TRP-channel family which is now known as TRPV family.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y5S1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51393
Name Human TRPV2 (aa 27-108) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias GRC; growth factor regulated channel; growth factor-regulated calcium channel; Osm-9-like TRP channel 2; OTRPC2; RP23-234K24.7; Sac2b; Stretch-activated channel 2 B; transient receptor potential cation channel subfamily V member 2; transient receptor potential cation channel, subfamily V, member 2; Trpv2; Vanilloid receptor-like protein 1; VRL; Vrl1; VRL-1
Common Name TRPV2
Gene Symbol Trpv2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RGKLDFGSGLPPMESQFQGEDRKFAPQIRVNLNYRKGTGASQPDPNRFDRDRLFNAVSRGVPEDLAGLPEYLSKTSKYLTDS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.