missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRPM7 (aa 1407-1549) Control Fragment Recombinant Protein

Product Code. 30209874
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209874

Brand: Invitrogen™ RP106377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111348 (PA5-111348. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96QT4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 54822
Name Human TRPM7 (aa 1407-1549) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4}
Common Name TRPM7
Gene Symbol Trpm7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SQPSCKSHLETGTKDQETVCSKATEGDNTEFGAFVGHRDSMDLQRFKETSNKIKILSNNNTSENTLKRVSSLAGFTDCHRTSIPVHSKQAEKISRRPSTEDTHEVDSKAALIPDWLQDRPSNREMPSEEGTLNGLTSPFKPAM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.