missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TRPM1 (aa 1348-1444) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (41%), Rat (41%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139905 (PA5-139905. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is similar to the transient receptor potential (Trp) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
Specifications
Specifications
| Accession Number | Q7Z4N2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4308 |
| Name | Human TRPM1 (aa 1348-1444) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4732499L03Rik; AI606771; CSNB1C; EMBL:CAI30141.1}; long transient receptor potential channel 1; Ltrpc1; melastatin; melastatin 1; melastatin-1; MLSN; MLSN1; transient receptor potential cation channel subfamily M member 1; transient receptor potential cation channel, subfamily M, member 1; transient receptor potential melastatin family; Trpm1; trpm1 {ECO:0000312 |
| Common Name | TRPM1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction