missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Troponin T (Cardiac) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB18741-SP
This item is not returnable.
View return policy
Description
Cardiac Troponin T Monoclonal specifically detects Cardiac Troponin T in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Cardiac Troponin T | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cardiac Muscle Troponin T, Cardiomyopathy, Dilated 1D (Autosomal Dominant), Cardiomyopathy, Hypertrophic, CMD1D, CMH2, CMPD2, cTnT, LVNC6, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), troponin T, cardiac muscle, Troponin T2, Cardiac Type, Truncated Cardiac Troponin T | |
| Human Cardiac Troponin T antigen Accession # P45379 | |
| 25 μg | |
| 7139 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry | |
| 990007 | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry 0.5-25 μg/mL | |
| P45379 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Troponin T(Cardiac) in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction