missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Troponin C ELISA Kit

Product Code. 17874150
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17874150

Brand: Invitrogen™ EH467RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Troponin C quantitates human Troponin C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Troponin C.

Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AI874626; cardiac 3; cardiac troponin C; Cardiac troponin I; CMD1FF; CMD1Z; CMD2A; CMH13; CMH7; cTnC; cTnI; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; Tn1; TnC; TN-C; tncc; Tni; TNN I3; TNNC; TNNC 1; TNNC1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; troponin C, slow skeletal and cardiac muscles; troponin C1, slow; troponin C1, slow skeletal and cardiac type; Troponin I; Troponin I cardiac; TroponinI
Assay Range 0.11 to 75 ng/mL
Assay Sensitivity 0.11 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7134
Gene Symbol Tnnc1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cardiac Troponin C
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.