missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TRMT11 (aa 149-228) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Packungsgröße:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (%), Rat (%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65067 (PA5-65067. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
Spezifikation
Spezifikation
| Accession Number | Q7Z4G4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 60487 |
| Name | Human TRMT11 (aa 149-228) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; putative RNA methylase MDS024; TRM11; Trmt11; TRMT11-1; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA guanosine-2'-O-methyltransferase TRM11-like protein-like protein; tRNA methyltransferase 11; tRNA methyltransferase 11 homolog; tRNA methyltransferase 11 homolog (S. cerevisiae) |
| Common Name | TRMT11 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur