missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TrkB Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF397
This item is not returnable.
View return policy
Description
TrkB Polyclonal specifically detects TrkB in Human samples. It is validated for Western Blot.
Specifications
| TrkB | |
| Polyclonal | |
| Biotin | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, NTRK2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4915 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NTRK2 | |
| Mouse myeloma cell line NS0-derived recombinant human TrkB | |
| 50 μg | |
| Primary | |
| Detects human TrkB in Western blots. In this format, less than 1% cross-reactivity with recombinant human (rh) TrkC, rhNT-3, rhNT-4, rhBDNF, and rh beta-NGF is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction