missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRIM27 (aa 136-231) Control Fragment Recombinant Protein

Product Code. 30180826
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180826

Brand: Invitrogen™ RP99567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nuclear matrix. It interacts with the enhancer of polycomb protein and represses gene transcription. It is also thought to be involved in the differentiation of male germ cells. Fusion of the N-terminus of this protein with the truncated C-terminus of the RET gene product has been shown to result in production of the ret transforming protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14373
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5987
Name Human TRIM27 (aa 136-231) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW538890; Gm19403; ret finger protein; ret finger protein 1; RFP; RFP transforming protein; Rfp1; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; RNF76; Trim27; tripartite motif containing 27; tripartite motif protein 27; tripartite motif protein TRIM27; tripartite motif-containing 27; tripartite motif-containing protein 27; Zinc finger protein RFP
Common Name TRIM27
Gene Symbol TRIM27
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VEGFKEQIQNQLDHLKRVKDLKKRRRAQGEQARAELLSLTQMEREKIVWEFEQLYHSLKEHEYRLLARLEELDLAIYNSINGAITQFSCNISHLSS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.