missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRIM26 (aa 60-123) Control Fragment Recombinant Protein

Product Code. 30182307
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182307

Brand: Invitrogen™ RP99610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62191 (PA5-62191. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12899
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7726
Name Human TRIM26 (aa 60-123) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Acid finger protein; AFP; AI462198; RING finger protein 95; RNF95; TRIM26; tripartite motif containing 26; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zfp173; Zfp1736; zinc finger protein 173; Znf173
Common Name TRIM26
Gene Symbol TRIM26
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FKKENIRPVWQLASLVENIERLKVDKGRQPGEVTREQQDAKLCERHREKLHYYCEDDGKLLCVM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.