missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TRIM25 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
TRIM25 Synthetic Peptide, PEP-0527, from Invitrogen.
TRIM25, also known as estrogen responsive finger protein (EFP) is a member of the RING finger-B box-coiled coil family and is a downstream target of estrogen receptor a. Mice lacking this gene displayed an underdeveloped uterus and reduced estrogen responsiveness demonstrating that this protein is essential for estrogen dependent proliferation. TRIM25 expression is increased in breast carcinoma where it promotes the growth of breast tumor by functioning as an E3 ubiquitin ligase that targets the negative cell cycle checkpoint 14-3-3s. Recent reports have indicated that TRIM25 E3 ubiquitin ligase is essential for the retinoic-acid-inducible gene 1 (RIG-1) signaling pathway to elicit host antiviral innate immunity.
Specifications
Specifications
| Gene ID (Entrez) | 7706 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q14258 |
| Common Name | Human TRIM25 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction