missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRIM21 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB62191
This item is not returnable.
View return policy
Description
TRIM21/SSA1 Monoclonal specifically detects TRIM21/SSA1 in Human samples. It is validated for Western Blot.
Specifications
| TRIM21/SSA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P19474 | |
| TRIM21 | |
| E. coli-derived recombinant human TRIM21 Arg195-Pro293 Accession # P19474 | |
| 100 μg | |
| Primary | |
| Detects human TRIM21 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TRIM5 or rhTRIM32 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 672714 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81, RNF81RING finger protein 81, ro(SS-A), Ro52, RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1, SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6737 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction