missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRIM Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4708N-100UG
This item is not returnable.
View return policy
Description
TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TRIM | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6PIZ9 | |
| TRAT1 | |
| E. coli-derived recombinant human TRIM Thr50-Asn186 Accession # Q6PIZ9 | |
| RUO | |
| 50852 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 488806 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRAT1, TRIMHSPC062 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TRIM in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction