missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRIF/TICAM1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB6216
This item is not returnable.
View return policy
Description
TRIF/TICAM1 Monoclonal specifically detects TRIF/TICAM1 in Human samples. It is validated for Western Blot.
Specifications
| TRIF/TICAM1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8IUC6 | |
| TICAM1 | |
| E. coli-derived recombinant human TRIF/TICAM1 Met474-Pro618 Accession # Q8IUC6 | |
| 100 μg | |
| Primary | |
| Detects human TRIF/TICAM1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) TRIF (aa 29-204), rhTRAM, rhTRIM, rhTRIM5, rhTRIM21, or rhTRIM32 is observed. | |
| Human | |
| IgG2a |
| Western Blot | |
| 567212 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM1, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 148022 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction