missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TREM2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB1828
This item is not returnable.
View return policy
Description
TREM2 Monoclonal specifically detects TREM2 in Human samples. It is validated for Western Blot.
Specifications
| TREM2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NZC2 | |
| TREM2 | |
| Mouse myeloma cell line NS0-derived recombinant human TREM-2 His19-Ser174 Accession # Q9NZC2 | |
| 100 μg | |
| Primary | |
| Detects human TREM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) TREM-1, rmTREM-2b, or rmTREM-3. | |
| Human | |
| Purified |
| Western Blot | |
| 263602 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54209 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction