missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TREM-2 ELISA Kit

Product Code. 17175249
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17175249

Brand: Invitrogen™ EH464RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human TREM-2 quantitates human TREM-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TREM-2.

TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; TREM2; TREM-2; Trem2a; Trem2b; Trem2c; triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2 A; triggering receptor expressed on myeloid cells 2 b; triggering receptor expressed on myeloid cells 2 c
Assay Range 28 to 7,000 pg/mL
Assay Sensitivity 28 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 54209
Gene Symbol Trem2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TREM2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.