missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Transthyretin Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Transthyretin Synthetic Peptide, PEP-0856, from Invitrogen.
Transthyretin (TTR), generally called prealbumin, is a plasma protein that plays an important role in physiology such as a transporter of hormone thyroxine and retinal-binding protein. After produced primarily in the liver, TTR is excreted into the plasma. TTR represents a disproportionate fraction (25%) of CSF protein, prompting the suggestion that it is either selectively transported across the blood-CSF barrier or synthesized de novo within the central nervous system. Transthyretin is a constituent found to the neuritic plaques, neurofibrillary tangles, and microangiopathic lesions of senile cerebral amyloid. It has been reported that more than 40 different mutations in the TTR gene associated with amyloid deposition.
Specifications
Specifications
| Gene ID (Entrez) | 7276 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P02766 |
| Common Name | Human Transthyretin |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction