Learn More
ACROBiosystems Human Transthyretin / Prealbumin Protein, His Tag
Description
Transthyretin (TTR) is also known as Prealbumin, ATTR, TBPA, PALB, which belongs to the transthyretin family. Transthyretin / TTR is a serum and cerebrospinal fluid carrier of the thyroid hormone thyroxine (T4) and retinol-binding protein bound to retinol. In cerebrospinal fluid TTR is the primary carrier of T4. TTR also acts as a carrier of retinol (vitamin A) through its association with retinol-binding protein (RBP) in the blood and the CSF. Less than 1% of TTR's T4 binding sites are occupied in blood. TTR misfolding and aggregation is known to be associated with the amyloid diseases.
Specifications
Specifications
| Accession Number | NP_000362 |
| Molecular Weight (g/mol) | 14.9 kDa |
| Quantity | 100 μg |
| Source | Human Transthyretin, His Tag (TTR-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Glu 147 (Accession # NP_000362). |
| Immunogen | Gly 21 - Glu 147 |
| Storage Requirements | -20°C |
| Gene Alias | Transthyretin,TTR,Prealbumin,TBPA,ATTR,PALB,CTS,CTS1,HsT2651 |
| Common Name | Transthyretin |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.