missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Transglutaminase 7/TGM7 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5426-SP
This item is not returnable.
View return policy
Description
Transglutaminase 7/TGM7 Monoclonal specifically detects Transglutaminase 7/TGM7 in Human samples. It is validated for Western Blot.
Specifications
| Transglutaminase 7/TGM7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q96PF1 | |
| TGM7 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 7/TGM7 Asp2-Pro710 Accession # Q96PF1 | |
| 25 μg | |
| Primary | |
| Detects human Transglutaminase 7/TGM7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human TGM2, 3, 4, or recombinant mouse TGM2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 602708 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| protein-glutamine gamma-glutamyltransferase Z, TG(Z), TGase Z, TGase-7, TGM7, TGMZ, TGMZEC 2.3.2.13, TGZ, transglutaminase 7, Transglutaminase Z, transglutaminase-7 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 116179 | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction