missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Transglutaminase 1/TGM1 (NP_000350) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Human Transglutaminase 1/TGM1 (NP_000350) VersaClone cDNA clone contains the complete ORF for Transglutaminase 1/TGM1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Transglutaminase 1/TGM1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
| Gene | hTGM1 |
| Accession Number | NP_000350 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl |
| Insert Type | 2466bp |
| pH | 8.5 |
| Gene ID (Entrez) | 7051 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction