missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAILR1/TNFRSF10A PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 19 publications
£230.00 - £431.00
Specifications
| Antigen | TRAILR1/TNFRSF10A |
|---|---|
| Clone | 69036 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18726687
|
R&D Systems
FAB347P-025 |
25 Tests |
£230.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776186
|
R&D Systems
FAB347P-100 |
100 Tests |
£431.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry.Specifications
| TRAILR1/TNFRSF10A | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TNFRSF10A, TRAIL R1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| TNFRSF10A | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects recombinant human TRAIL R1/TNFRSF10A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TRAIL R2, rhTRAIL R3, rhTRAIL R4, or rhDcR3 is observed. |
| 69036 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| AAC51226 | |
| 8797 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TRAIL R1/TNFRSF10A Ala24-Asn239 Accession # AAC51226 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title