missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TRAIL Receptor-1 (soluble) Recombinant Protein, PeproTech®

Product Code. 17892183
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17892183

Brand: Gibco™ 31018100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human soluble TRAIL Receptor-1/DR4 is a 22.7 kDa protein (215 amino acid residues) consisting of the TNFR homologous, cysteine-rich portion of the extracellular domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00220
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 8797
Molecular Weight (g/mol) 22.7 kDa
Name Human TRAIL Receptor-1 (soluble)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias APO2; CD261; cytotoxic TRAIL receptor; death receptor 4; DR4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF receptor superfamily member 10 A; TNF-related apoptosis-inducing ligand receptor 1; TNFRSF10A; TRAIL R1; TRAIL receptor 1; TRAILR1; TRAIL-R1; TRAILR-1; tumor necrosis factor receptor superfamily member 10 A; tumor necrosis factor receptor superfamily member 10 A variant 2; tumor necrosis factor receptor superfamily, member 10 A
Common Name DR4
Gene Symbol TNFRSF10A
Biological Activity Measured by its ability to inhibit apoptosis in LN-18 cells. The expected ED50 for this effect is 0.4 -0.5 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SGTGAAAATP SKVWGSSAGR IEPRGGGRGA LPTSMGQHGP SARARAGRAP GPRPAREASP RLRVHKTFKF VVVGVLLQVV PSSAATIKLH DQSIGTQQWE HSPLGELCPP GSHRSERPGA CNRCTEGVGY TNASQQLFAC LPCTACKSDE EERSPCTTTR NTACQCKPGT FRNDNSAEMC RKCSTGCPRG MVKVKDCTPW SDIECVHKES GNGHN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.