missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAIL R3/TNFRSF10C PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: R&D Systems FAB6302P
This item is not returnable.
View return policy
Description
TRAILR3/TNFRSF10C Monoclonal specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Flow Cytometry.
Specifications
| TRAILR3/TNFRSF10C | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAILR3, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8794 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 90906 | |
| Flow Cytometry 10 uL/10^6 cells | |
| O14798 | |
| TNFRSF10C | |
| Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C Met1-Ala221 Accession # O14798 | |
| 100 Tests | |
| Primary | |
| Detects human TRAIL R3/TNFRSF10C in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) TRAIL R2, rhTRAIL R1, or rhTRAIL R4 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction