missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRAIL R2/TNFRSF10B (NP_003833) VersaClone cDNA

Product Code. 18759658
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18759658 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759658 Supplier R&D Systems™ Supplier No. RDC1434
  • ONLINE EXCLUSIVE PRICE
    £301.00
    £284.55 / 10µg
    Save £16.45
    5% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human TRAIL R2/TNFRSF10B (NP_003833) VersaClone cDNA clone contains the complete ORF for TRAILR2/TNFRSF10B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TRAILR2/TNFRSF10B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Gene TRAIL R2/TNFRSF10B
Accession Number NP_003833
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.107
Insert Type 1336bp
Gene ID (Entrez) 8795
Gene Alias CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Show More Show Less

For Reasearch Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.