missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRAF5 Control Fragment Recombinant Protein

Product Code. 30202980
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202980

Brand: Invitrogen™ RP89647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52507 (PA5-52507. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e. g. RANK, CD30, CD40, etc. ) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6) (1-4). TRAF5 is a 558-amino acid protein. TRAF5 is implicated in NF-kB and c-Jun NH(2)-terminal kinase/stress-activated protein kinase activation by members of the TNF receptor superfamily, including CD27, CD30, CD40, and lymphotoxin-beta receptor. Targeted disruption of TRAF5 gene causes defects in CD40-CD27 mediated lymphocyte activation (5).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00463
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7188
Name Human TRAF5 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2700049M22Rik; AI875481; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; MGC:39780; p11 BID; p13 BID; p15 BID; p22 BID; RING finger protein 84; RNF84; TNF receptor associated factor 5; TNF receptor-associated factor 5; Traf5
Common Name TRAF5
Gene Symbol TRAF5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NAKVILGRYQQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.