missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAF-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3284-SP
This item is not returnable.
View return policy
Description
TRAF-6 Monoclonal specifically detects TRAF-6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| TRAF-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6, TRAF6 | |
| E.coli-derived recombinant human TRAF-6 Met1-Val522 Accession # Q9Y4K3 | |
| 25 μg | |
| Primary | |
| Detects human TRAF-6 in direct ELISAs. | |
| Human | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 326019 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9Y4K3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7189 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction