missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAF-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3279-SP
This item is not returnable.
View return policy
Description
TRAF-4 Monoclonal specifically detects TRAF-4 in Human samples. It is validated for Western Blot.
Specifications
| TRAF-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2.5 ug/mL | |
| Q9BUZ4 | |
| TRAF4 | |
| E. coli-derived recombinant human TRAF-4 Met1-Ser470 Accession # Q9BUZ4 | |
| 25 μg | |
| Primary | |
| Detects endogenous human TRAF-4. In Western blots, this antibody does not cross-react with rhTRAF-1, -2, -3, -5, or -6 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 433709 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CART1, CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, TRAF4, tumor necrosis receptor-associated factor 4A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9618 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction