missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRAF-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3276-SP
This item is not returnable.
View return policy
Description
TRAF-1 Polyclonal specifically detects TRAF-1 in Human samples. It is validated for Western Blot.
Specifications
| TRAF-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EBI6, EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1, TRAF1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7185 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q13077 | |
| TRAF1 | |
| E. coli-derived recombinant human TRAF-1 Met1-Thr416 Accession # Q13077 | |
| 25 μg | |
| Primary | |
| Detects human TRAF-1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human TRAF-2, -3, -4, -5, and -6 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction