missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TRACP/PAP/ACP5 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human TRACP/PAP/ACP5 quantitates human TRACP/PAP/ACP5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TRACP/PAP/ACP5.
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
Specifications
Specifications
| Accession Number | acid phosphatase 5, tartrate resistant; acid phosphatase type V; ACP5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5 A; tartrate-resistant acid phosphatase 5 b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; Trap; TR-AP; TRAPase; TrATPase; TTRRAP; type 5 acid phosphatase |
| Assay Range | 0.05 to 10 ng/mL |
| Assay Sensitivity | 0.05 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 54 |
| Gene Symbol | ACP5 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction