missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRACP/PAP/ACP5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £381.00
Specifications
| Antigen | TRACP/PAP/ACP5 |
|---|---|
| Clone | 417603 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778339
![]() |
R&D Systems
MAB3948-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152101
|
R&D Systems
MAB3948 |
100 μg |
£381.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRACP/PAP/ACP5 Monoclonal specifically detects TRACP/PAP/ACP5 in Human samples. It is validated for Western Blot.Specifications
| TRACP/PAP/ACP5 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P13686 | |
| 54 | |
| Mouse myeloma cell line NS0-derived recombinant human TRACP/PAP/ACP5 Ala22-Pro320 Accession # P13686 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TRACP in direct ELISAs and Western blots. |
| 417603 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACP5, HPAP, TRACP5a, TRACP5b, TRACP-5b, TRAP, TrATPase, TTRRAP | |
| ACP5 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title